Human Cancer Cell Line Blot, G-Biosciences
Catalog #: CA82023-240
Supplier: G-Biosciences
Specifications
- Description:Human Cancer Cell Line Blot
- Blot lane order:1. Protein marker
2. Acute promyelocytic leukemia (HL-60)
3. T lymphocyte lymphoma (HUT-78)
4. B cell lymphoma (BJAB)
5. Mammary adenocarcinoma (MCF7)
6. Amelanotic melanoma (C32)
7. Acute T cell leukemia (JURKAT)
8. Chronic Myelogenous leukemia (K562)
9. Epidermoid carcinoma (A-431)
10. Epithlioid carcinoma (HeLa)
11. Burkitt's lymphoma (Ramos)
12. Prostate adenocarcinoma (LNCap)
13. Hepatoblastoma (HepG2)
14. Neuroepithelioma (SK-N-MC) - Source:Human
- Tissue:-
- Type:—
- Supplier No.:TB55
Specifications
About this item
G-Biosciences offers ready-to-screen, cancerous tissue human cell line blots.
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information on the blots as G-Biosciences reserves the right to change the blot profile due to the availability of GenLysate™.
Ordering information: Accessories available.
Customers Who Bought This Also Bought