Sub-Cellular Fraction Specific Blot, G-Biosciences
Catalog #: CA82023-224
Supplier: G-Biosciences
Specifications
- Description:Mouse Sub-Cellular Fraction Specific Blot
- Blot lane order:1. Protein marker
2. Mouse Whole Liver
3. Mouse Nuclei Lysate
4. Mouse Mitochondria Lysate
5. Mouse Golgi Complex Lysate
6. Mouse Smooth Endoplasmic Reticulum Lysate
7. Mouse Rough Endoplasmic Reticulum Lysate - Cat. no.:CA82023-224
- Source:Mouse
- Tissue:-
- Supplier No.:TB47
Specifications
About this item
G-Biosciences offers ready-to-screen, subcellular fractions from liver tissue, prepared by carefully dissecting out anatomically and functionally distinct regions of adult mouse livers.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information on the blots as G-Biosciences reserves the right to change the blot profile due to the availability of GenLysate™.
Ordering information: Accessories available.
Customers Who Bought This Also Bought