Modern Round Bottles, Nylon/PE, Narrow Mouth, Qorpak®
Supplier: Qorpak
Certificates
About this item
Nylon/PE barrier modern round bottles are nylon bottles co-extruded with PE & banded together.
This barrier prevents trace ingredients getting into the 'sidewall maze'. Containers produced by this process offer improved protection for non-polar hydrocarbons.
Customers Who Bought This Also Bought