Boston Bottles, Round, Low-Density Polyethylene, Narrow Mouth, Qorpak®
Supplier: Qorpak
Certificates
About this item
Natural LDPE, round bottles are the perfect solution for liquid sample handling in a squeezable bottle.
These bottles are virtually unbreakable and more resistant to stress cracking than HDPE bottles. They retain their impact strength at very low temperatures.
Specifications
- Catalog No:
- 16054-006
- 76311-220
- 16054-002
- 16054-003
- 16054-004
- 16054-005
- 16218-144
- 16807-132
- 16087-134
- 16085-001
- 75856-978
- Material:
- LDPE
- LDPE
- LDPE
- LDPE
- LDPE
- LDPE
- LDPE
- LDPE
- LDPE
- LDPE
- LDPE
- Colour:
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque