Specifications
- Description:Genomic Tube-O-DIALYZER™
- Size:25 Preps
- Cat. no.:CA82022-546
- Supplier No.:786-142-45MC
Specifications
About this item
G-Biosciences' Genomic Tube-O-DIALYZER™ allows for rapid dialysis of small genomic DNA samples (>100kb) to remove small fragment DNA, salts, RNA and other contaminants with minimal hands-on manipulation
G-Biosciences' Genomic Tube-O-DIALYZER™ allows for 100% sample recovery. There is no left over sample as in a dialysis bag.
G-Biosciences' Genomic Tube-O-DIALYZER™ is supplied with 0.45 µm size membrane, which allows rapid removal of impurities, RNA, small fragment DNA, etc. and retains only genomic DNA.
G-Biosciences' Genomic Tube-O-DIALYZER™ kit is supplied with 25 genomic dialyzer tubes, 25 caps, 6 floats, and a pair of forceps. Each tube can hold a 0.2 to 2.5 ml sample.
The simple, patented design of the Genomic Tube-O-DIALYZER™ includes a single tube for dialysis and storage. This ensures no sample loss and no contamination.