About this item
This universal well designed meter is the perfect economical choice for a wide range of applications.
- Universal well design
- Multiparameter
- Includes electrode holder
- Faster measurements
- Color display
Not only does it measure conductivity but also various other parameters such as salinity, resistivity, total dissolved solids and conductivity ash.
The included electrode holder with the perfectly vertical uPlace™ movement helps you bring your electrode in the position where it works best for your sample. This allows for faster measurements and reduces the risk of the sample vessel tipping over and/or damage the sensor.
SevenCompact™ gives you the choice to view all measurement information on the display or to focus on the essentials with the uFocus™ view. Switching between the different display layouts is easy, requiring only a single keypress. The new color display with its well-designed icons and menu settings in 10 languages makes operation genuinely intuitive. Applications range from routine measurements through sample analysis, data handling and data archiving in compliance with GLP regulations. The meters have a simple, innovative design that meets the requirements for a universal, easy-to-operate conductivity meter.