Nalgene® Stainless Steel Storage Racks for Microplates, Thermo Scientific
Catalog #: 62409-082
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:80 Low profile, 48 Standard, 16 Deep well
- Cat. no.:62409-082
- No. of Places:4 x 4
- L×W×H:38.7 x 13.9 x 21.6 cm (15 x 51/2 x 81/2")
- Supplier No.:5039-0048
Specifications
About this item
Nalgene® Stainless Steel Storage racks for Micro plates come in two convenient horizontal sizes to fit your freezer or refrigerator
These storage racks have sturdy handles on top and on the sides for easy transport. Each compartment on the racks comes with a retainer clip which holds the plates securely in place.
Customers Who Bought This Also Bought