Eppendorf® MasterClear™ Cap Strips and real-time PCR Tube Strips
Supplier: Eppendorf
Certificates
About this item
The Masterclear™ Cap Strips offer distinct advantages for real-time PCR applications.
The real-time PCR Tube Strips feature an extremely thin wall for optimal heat transfer. Designed to provide high mechanical stability, they are ideal for real-time PCR applications, especially for use with small reaction volumes. White wells offer improved reflection.
The inverted dome of the cap strips prevents scratching or contamination of the optical surface and reduces the volume of the micro test tube. Additionally, the material and the production processes were optimized to ensure excellent light transmission.
Specifications
- Catalog No:
- CA89094-210
- CA89094-212
- CA89094-214
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001