BLOK™ BSA, 10% BSA Protein Blocking Solution, G-Biosciences
Catalog #: CA82022-632
Supplier: G-Biosciences
CAS Number:
Specifications
- Description:BLOK™ BSA in TBS, 10% solution
- Size:125 ml
- Cat. no.:CA82022-632
- Application:10% BSA Solution for Blocking Western, ELISA, and Dot Blots
- Supplier No.:786-193
Specifications
About this item
G-Biosciences' BLOK™ BSA is a 10% BSA protein blocking solution suitable for blocking Western blots, ELISA and dot Blots
G-Biosciences' BLOK™ BSA in TBS and in PBS do not contain endogenous biotin; therefore, they are compatible with biotin/avidin systems and result in high signal-to-noise ratios.
Supplied in ready-to-use form, G-Biosciences offers BLOK™ BSA in your choice of TBS or PBS buffers.
Ordering information: Accessories available.
Customers Who Bought This Also Bought