Rabbit Surgery Board, Plas-Labs™
Supplier: Plas-Labs
Certificates
About this item
Surgery boards are formed one piece high-temp Noryl™ for durability and ease of cleaning
Adjustable nylon cords are included.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.