Rodent Animal Restrainer, Plas-Labs™
Supplier: Plas-Labs
Certificates
About this item
The clear acrylic cylinder has an open groove through which the animal's tail is drawn
Do not Autoclave or Sterilize with Acetone.
May be chemically sterilized or washed in temperatures up to 82.2°C.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.