About this item
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. Super Syringes are large volume Gastight® syringe. Super Syringes are the only Hamilton syringes made with an acrylic barrel instead of a glass barrel. They are suitable for applications such as air and gas collection in industrial laboratories, aid for the graduation of pneumatographs and small containers and pulmonary function tests. Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze. These syringes are ideal for dispensing volumes from 50 mL up to 2 L. A variety of different terminations and needle options are available.
Trachael- This termination accepts 5/8 inch inner diameter flexible tubing. PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. The version with slots is used during the attachment to some instruments but does not effect the connection to needles. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
Point styles are available on Kel-f Hub and Metal Hub Needles. Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34-22. Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34-22. Point Style 4 - Standard 12° beveled needle point is recommended for life science applications. Available gauges: 34-22. Special point styles such as 30°, 45°, or any other angle are available upon request. Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10. Point Style AS - Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration. Available gauges: 26-10.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 72310-298
- 60377-040
- 72310-300
- 72310-294
- 89221-478
- Model:
- S1000 TLL Syringe
- S1000 Tracheal Syringe
- S0500 TLL Syringe
- S2000 TLL Syringe
- Type:
- GASTIGHT
- GASTIGHT
- GASTIGHT
- GASTIGHT
- Series:
- Super Syringe
- Super Syringe
- Super Syringe
- Super Syringe
- Fluid path:
- Buna-N, Nickel plated brass, PTFE, Silicone
- Buna-N, Nickel plated brass, PTFE, Silicone
- Buna-N, Nickel plated brass, PTFE, Silicone
- Buna-N, Nickel plated brass, PTFE, Silicone
- Max. pressure:
- 30 psig (2.1 bar)
- 30 psig (2.1 bar)
- 30 psig (2.1 bar)
- 30 psig (2.1 bar)
- Max. temperature:
- 80 °C / 176 °F
- 80 °C / 176 °F
- 80 °C / 176 °F
- 80 °C / 176 °F
- Min. temperature:
- 0 °C / 32 °F
- 0 °C / 32 °F
- 0 °C / 32 °F
- 0 °C / 32 °F