VITLAB® Erlenmeyer Flasks with Screw Caps, PP, BrandTech
Supplier: Brandtech
Certificates
About this item
Flasks are break-resistant, autoclavable, and easy to clean.
- Wide-mouth design for easy pouring
- Printed scale of approximate volumes
- Made of translucent polypropylene
Compatible with ground joint stoppers (not included).
Ordering information: Supplied with a polypropylene screw cap.
Customers Who Bought This Also Bought