Order Entry
Canada
Orders LinkContactUsLinkComponent
Immobilized Trypsin

Immobilized Trypsin

2 ml resin
Catalog # CA89168-040
Supplier:   G-Biosciences
Supplier #:  786-792
Immobilized Trypsin
Catalog # CA89168-040
Supplier:  G-Biosciences
CAS Number:  
Immobilized Trypsin

2 ml resin

CAS Number:  
Price
$437.64
Log in for availability
Each (2ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Immobilized Trypsin is TPCK Treated Trypsin immobilized on 4% agarose that eliminates the contamination of protein digests by the trypsin

Trypsin is a serine endopeptidase that specifically cleaves peptide bonds on the carboxy side of s-aminoethyl cysteine, arginine and lysine residues and typically there is little or no cleavage at arginyl-proline and lysyl-proline bonds. The distribution of these residues in proteins allows trypsin digestion to produce peptides that are readily identified by mass spectrometry. Native trypsin is prone to autolysis that results in pseudotrypsin, which exhibits a broader proteolytic specificity (a chymotrypsin like activity) and trypsin fragments that interfere with sequence analysis.

The Trypsin is TPCK treated to inactive the interfering chymotrypsin activity and the resulting protein is affinity purified. Immobilzed Trypsin is supplied as a 50% slurry containing glycerol and sodium azide as a preservative.

The immobilized trypsin is readily removed by separating the agarose from the digestion solution.

Ordering information: Accessories not included.

Specifications

Specifications

  • Size:
    2 ml resin
  • Cat. No.:
    CA89168-040
  • Enzyme name:
    Immobilized Trypsin

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.