uTIP™ Filtered Pipette Tips for Universal Pipettors, Biotix
Supplier: Biotix
Certificates
About this item
Biotix filtered tips provide protection against aerosol contamination (cross contamination) with a 99.99% bacterial filter efficiency (BFE). uTIP is compatible with major brands like Eppendorf, Gilson, Biohit and Thermofisher.
- FlexFit® technology features a flexible proximal end to ensure a secure seal with less force then competitor tips
- X-Resin® is our proprietary low retention resin which maximizes sample recovery and improves accuracy and precision
- Blade® Technology makes a fine point at the distal end of the tip ensuring a full dispense without the need for tip touch off
Each lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled and inspected to ensure only the best go into our products. All racks include a label with a unique bar-coded serial number, part number, and description for traceability.
Related Information
Certificates
Search by Lot Number76175-406_241625
Conformity declaration
89136-568
Images 360
Biotix Blade Filter Pipets Demonstration Biotix Delta Filter Demonstration Biotix FlexFit Features Biotix X-Resin Overview
Videos
Biotix uTIP Pipette Tips Compatible With Universal Pipettes VWR® Balances
Product Literature
Customers Who Bought This Also Bought