Specifications
- Size:Each
- Enzyme name:ExiSpin
- Cat. no.:89170-878
- Supplier No.:A-7040
Specifications
About this item
ExiSpin has the functions of a vortexer as well as a microcentrifuge and will significantly save you time in the lab
The time and strength of centrifugation/mixing is fully programmable. The numbers of spin-mix-spin cycles can be set (1-999). After this input, a single button automatically starts the process as follows:
It provides reproducible results using protocols optimized to samples by preventing the possibilities of experimental errors. Four 8-strips tubes can be handled simultaneously, which enable 32 PCR/QPCR mixture tubes ready to run at once. Twelve 1.5 ml tubes can be loaded for simultaneous cell lysis and subsequent centrifuge-down. The programmed speed and time quarantine the reproducibility of each application.
ExiSpin comes complete with rotors for 4 x 8-strip tubes and 12 x 1.5/2.0 ml tubes. Use it for Minipreps, to resuspend oligos, set up you PCR reactions, or any task that requires vortexing in micro test tube or 0.2 ml PCR tubes.