VWR® READY-PACK™ Ultra Pure Grade TRIS Buffered Saline (TBS)
Catalog #: 97062-370
Supplier: VWR
CAS Number:
Specifications
- Size:2 Packs
- Cat. no.:97062-370
- Supplier No.:0788-2PK
Specifications
About this item
Pre-blended TBS buffer
Each Ready-Pack™ prepares 1 liter of a 20X solution of tris buffered saline when dissolved in water. A 1X solution contains 25mM Tris, 140mM Sodium Chloride, and 3.0mM Potassium Chloride.
Customers Who Bought This Also Bought