Nalgene® Low-Density Polyethelene Low Particulate and Low Metals Bottles, Thermo Scientific
Catalog #: 89091-772
Supplier: Thermo Fisher Scientific
Specifications
- Cap Size:24 mm
- Capacity:250 ml (8.5 oz.)
- Ø:60 mm
- Cat. no.:89091-772
- Supplier No.:382003-0008
- Height with Cap:134 mm
- Height without Cap:131 mm
Specifications
About this item
Nalgene® low-density polyethelene low particulate and low metals bottles are narrow-mouth bottles with particle levels of less than 20 particles per ml at 0.3 µm and greater.
Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. The bottles are excellent for ICP-MS reagent and standard storage and include a certificate of quality that assures the products have been tested and accepted in accordance with specifications.
Certifications: Metals certified to (µg/L) ppb levels <0.20 Hg, <0.5 Be, <1.0 As, Cd, Pb, <2.0 Sb, Se, <5.0 Ag, Co, Cr, Cu, Mn, Th, V, <10 Ba, Ni, Zn, <50 Mg, <75 Al, <100 Ca, Fe, K, Na.
Customers Who Bought This Also Bought