Closures for RSA™ Screw-Thread Vials, Amber, MicroSolv
Catalog #: 97055-428
Supplier: MicroSolv Technology Corp
Specifications
- Cap Size:9-425 mm
- Cap Type:Screw cap
- Septum:Fitted Silicone Rubber/PTFE, ultra-pure
- Material:PP
- Cat. no.:97055-428
- Cap Color:Light blue
- Supplier No.:9509S-10C-B