About this item
This system consists of a hematology analyzer, centrifuge, and external printer. The QBC Autoread Plus Dry Hematology System is a reliable and rugged complete blood count analyzer used by thousands of practitioners around the globe. A cost-effective and virtually maintenance free system, the Autoread Plus provides point-of-care CBCs in just 7 minutes.
The centrifuge uses QBC® AccuTubes, which hold a 70µL sample size of capillary or venous blood. They feature pre-inserted ventilated stoppers and external floats.
This system consists of a hematology analyzer, centrifuge, and external printer. The QBC Autoread Plus Dry Hematology System is a reliable and rugged complete blood count analyzer used by thousands of practitioners around the globe. A cost-effective and virtually maintenance free system, the Autoread Plus provides point-of-care CBCs in just 7 minutes.
The centrifuge uses QBC® AccuTubes, which hold a 70µL sample size of capillary or venous blood. They feature pre-inserted ventilated stoppers and external floats.
The hematology analyzer measures nine important CBC parameters from venous or capillary blood samples. It uses dry hematology technology instead of liquid reagents to save storage space, cut costs, and reduce waste. The system is easy to transport, but its hard plastic frame is durable in the harshest conditions. Samples are centrifuged in five minutes and results automatically print when ready. The system is permanently calibrated during manufacturing
AccuTubes contain acridine orange dye required to stain blood samples for analysis in the Autoread Plus. AccuTubes can be used for capillary or venous blood samples using the pipetter. They are packaged with a ventilated stopper at the end of the tube, and soft grip float holder for easy insertion of the plastic float.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89031-550
- 89031-548
- Sample volume:
- 70 L (AccuTube), 60 L (Standard Capillary/E-Z Prep Capillary)
- Operating temperature:
- 20 to 37°C (68 to 98°F)
- Electrical:
- 90–265 V; 50–60 Hz
- Dimensions:
- 34.3H x 24.1W x 10.2D cm (13½ x 9½ x 4”)
- Weight:
- 3.6 kg (8 lbs.)