Dermal Curettes, Integra™ Miltex®
Supplier: Miltex
Certificates
About this item
Curettes feature a seamless, stainless steel semi-sharp cutting edge with a unique symmetrically ground beveled edge, and a textured handle for better comfort and control
Sterile and disposable.
Ordering information: Curettes are individually packaged in peel-apart pouches.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.