Specifications
- Description:Transport Kit
- Cat. no.:89171-032
- Supplier No.:37939
Specifications
About this item
Transport kit features all the necessary collection tools for pathology testing
To ensure safe transport, all supplies are placed into appropriate cushioning material with an absorbent pad and a secondary leak-resistant bag. The kit can be placed inside a carrier shipping envelope with attached air waybill. Kit includes two pre-filled formalin containers, slide mailer with five 75x25mm (3x1”) single frosted-end slides, one 6mL EDTA tube, and two 4mL sodium heparin tubes