To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:ExiPrep™ 16 Plus
- Cat. no.:89170-874
- Supplier No.:A-5030
Specifications
About this item
These laboratory automated systems are able to extract DNA or RNA from various biological samples including tissue, cell culture, whole blood, serum, and plasma in just 40–60 minutes
The ExiPrep 16 Pro has TCO/IP network capability to allow multiple units to operate in parallel. It also features a built-in cooling block that can keep samples at 10°C for as long as necessary.
They are also able to purify His-tag recombinant proteins. Both the ExiPrep™ 16 Plus and ExiPrep™ Pro can handle up to 16 samples at once, and include optimized extraction protocols for many sample types. Intuitive touch screen interfaces enable simple operation. The automatic UV sterilization lamps and unique contamination shields that slide underneath pipet tips eliminate the possibility of intra-assay cross-contamination. Integrated combined magnetic/heating blocks reduce experiment time and increase elution efficiency
Specifications
Electrical: 110–220V, 50/60Hz, 2.7A
W×H×D: 320 x 487 x 535 mm
Weight: 22 kg (48.5 lbs.)