Specifications
- Size:4.4 g
- Cat. no.:CACMB41363
- Supplier No.:402
Specifications
About this item
Each vial contains enough buffer to prepare one gallon of pH 7.15 buffer solution for use in the five-step staining process
Each vial contains enough buffer to prepare one gallon of pH 7.15 buffer solution for use in the five-step staining process
The buffer is helpful with the development of the correct, clear nuclear color schemes.