Epredia™ Superfrost™ Excell™ Microscope Adhesion Slides, Thermo Scientific
Supplier: Erie Scientific
Certificates
About this item
Slide surface has superior adhesion and wettability
Slide surface has superior adhesion and wettability
Developed for use in HIER methods requiring high pH antigen retrieval solutions, including EDTA.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.