ContactUsLinkComponent
BD Q-Syte™ Closed Luer Access Split Septum Device, BD Medical
BD Q-Syte™ Closed Luer Access Split Septum Device, BD Medical
Catalog #: CABD385100
Supplier:  BD
BD Q-Syte* Luer Access Device
BD Q-Syte™ Closed Luer Access Split Septum Device, BD Medical
Catalog #: CABD385100
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Split Septum Device
  • Supplier No.:
    385100
Specifications
About this item
Split septum device is designed to reduce catheter-related bloodstream infections associated with needleless IV systems
The smooth surface is easy to clean because there are no crevices or gaps to house bacteria. The simple fluid path design reduces places for microbes to grow and delivers a better flow rate. Clear housing allows for visual assessment of the fluid path. The Acacia Extension Set includes a 15cm (6”) small bore bi-extension and spin nut.
Customers Who Bought This Also Bought