Specifications
- Description:Woll Drain Dissecting Tray
- Cat. no.:15640-516
- Supplier No.:360
Specifications
About this item
Designed to restrict contamination by directing spillage into a sink
Useful for cleaning glassware, urinalysis, solution preparation, bacterial preparation, and other lab applications. Spout at the end of the tray is 12.7Wx7.6Lcm (5x3") with a 20˚ slope. Adjustable tray slope.