About this item
High temperature adhesive for bonding ceramic to metal.
Mix ratio (w/w): 1.5:1 powder to liquid.
Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C
UN No.: 3253 MDL No.: MFCD00011109
High temperature adhesive for bonding ceramic to metal