Manganese(II) chloride, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Synonyms: Manganous chloride
Formula:MnCl₂
MW:125.84 g/mol
Boiling Pt:1190 °C
Melting Pt:650 °C
Density:2.977
MDL Number:MFCD00011114
CAS Number:7773-01-5
ADR:6.1,III
Merck Index:12,05768
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,99
:Boiling Pt:
:Boiling Pt: 1190 °C
:Melting Pt:
:Melting Pt: 650 °C
:Flash Pt:
:Flash Pt:
:M.W. 125.84 g/mol
:Manganese(II) chloride
Storage Temperature:Ambient
EINECS:231-869-6
:1190 °C
UN:3288
:6.1
:III
:GHS06
:GHS09
:14,5728
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Manganese(II) chloride, ultra dry ≥99.99% (metals basis):
Specification Test Results
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Alfa-Aesar High-Purity Inorganics
Product Literature
Ultra Dry Brand Description.jpg
Datasheet
Customers Who Bought This Also Bought