About this item
The Hoshizaki produces small cubelet nugget-style ice and water dispenser features either LED touch buttons (89166-544) or Opti-Serve touchless operation (76808-992). Both models feature a durable stainless steel exterior, advanced CleanCycle24™ design, and stainless steel auger with greaseless bearing.
- Up to 618 lbs. of ice produced per 24 hours
- Up to 40 lbs. of built-in storage capacity (1.3 ft.³)
- Place on countertop or optional stand: SD-450, SD-500
- Protected by H-GUARD Plus antimicrobial agent
- R-404A refrigerant (1 lbs. 4.1 oz./570 g)
- Electrical: 115 V, 60 Hz, 1 phase
- Power: 12.9 A, 5.62 kW
The 76808-992 and 89166-544 are capable of producing approximately 618 lbs. of cubelet nugget-style ice cubes per day. The built-in storage bin will hold up to 40 lbs. of ice* at a time. This Air-cooled icemaker has an LED remote sensor for hands-free operation with opaque containers. This sanitary icemaker/dispenser features a user-friendly dispensing mechanism that allows easy to chew, cubelet ice to be dispensed with a simple push of a button. This model features CleanCycle12™ – every 12 h, it performs 15 min purge to clean itself and it is protected by H-GUARD plus antimicrobial agent. The sanitary dispenser design keeps ice fully contained until it is needed, minimizing the opportunity for cross-contamination and keeping the ice clean. Units are hardwired.
It has LED remote sensor for hands-free operation with opaque containers.