PYREX® 3-Hole Mobile Phase Delivery Cap, Corning
Catalog #: 71001-620
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Description:PYREX® 3-Hole Mobile Phase Delivery Cap
- Cat. no.:71001-620
- Supplier No.:1395-3H
Specifications
About this item
The delivery cap top has three holes that have 1/4" - 28 threads with 1/8" O.D
For use with PYREX® HPLC Reservoirs, Single Cavity.
PTFE tubing. The body is glass-filled PTFE (Polytetrafluoroethylene). PET (Polyethylene terephthalate) screw cap has a GL-45 thread. The cap includes three 1/4" - 28 ETFE (Ethylene tetrafluoroethylene) plugs and a Viton® O-ring for sealing to the reservoir. Integral pressure relief valve relieves reservoir pressure at 10 psi.
Customers Who Bought This Also Bought