PYREX® Knob Top Desiccators, Corning
Supplier: Corning
Certificates
About this item
These PYREX® borosilicate glass desiccators are designed for general analytical work. Material is Borosilicate Glass Code 7740.
- Sturdy knobs make the covers easy to handle
Ordering information: Plate is not included. For a compatible plate, please contact your representative for more information.
Caution: Warning: Do not heat or subject to pressure. Not suitable for vacuum applications.
Specifications
- Catalog No:
- 89134-400
- 89134-384
- 89134-386
- 89134-390
- 89134-388
- 89134-394
- 89134-392
- 89134-398
- 89134-396
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing