About this item
HAEMO-SOL® Regular is a concentrated all- purpose cleaner formulated to thoroughly clean and remove soils from laboratory and medical instruments.
- No caustic chemicals or odor
- Leaves no residue, spots of film after rinsing
Mix 1/2 ounce to 1 gallon of hot water. Soak, agitiate and rinse items thoroughly. One jar produces 160 gallons of cleaning solution.
Designed for use in soak and rinse cleaning, as well as ultrasonic cleaning.