About this item
Pads are used to hold biopsies in place during processing
Biopsy samples are sandwiched between two foam pads and are placed either in tissue capsule or cassettes with metal or plastic lids. Specially designed polyester urethane foam achieves optimum solvent flow
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot Number60872-492_905091396 60872-492_910091836 60872-492_913182346