UniMailer™ Slide Mailers, Simport Scientific
Supplier: Simport Scientific
Certificates
About this item
These one-slide trays are stackable and are secured by a unique locking mechanism
These one-slide trays are stackable and are secured by a unique locking mechanism
They are designed to accommodate standard slides with or without cover glasses. Mailers hold slides horizontally and are designed for an exact fit to prevent breaking, but slides are easily removed as needed
Temperature range: –80 to 80°C
Customers Who Bought This Also Bought