2-Propanol ≥99% for biotechnology
Catalog #: 97064-700
Supplier: VWR
CAS Number: 67-63-0
Specifications
- Size:1 L
- Supplier No.:0918-1L
Specifications
Specification Test Results
- Color (APHA):= 10
- Density:0.782 g/mL - 0.788 g/mL
- Moisture (KF):= 0.2 %
- Purity:= 99 %
- 2-Propanol ≥99% for biotechnology:
Specification Test Results
About this item
Synonyms: IPA,Isopropyl Rubbing Alcohol,Isopropyl alcohol,Dimethyl carbinol,2-Hydroxy propane,Isopropanol
Boiling Pt:82 °C
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
Flash Pt:12 °C
:Boiling Pt:
:Boiling Pt: 82 °C
:Melting Pt:
:Melting Pt: –89 °C
:Flash Pt:
:Flash Pt: 12 °C
:M.W. 60.1 g/mol
Merck Index:13,05228
:2-Propanol
Formula:(CH₃)₂CHOH
MW:60.1 g/mol
CAS Number:67-63-0
EINECS:200-661-7
:82 °C
UN:1219
REACH:01-2119457558-25
:3
:II
:GHS07
:GHS02
Storage Temperature:Ambient
MDL Number:MFCD00011674
Customers Who Bought This Also Bought