Water for biotechnology nuclease-free, 0.2 µm filtered
Supplier: VWR
Certificates
About this item
Synonyms: Hydrogen oxide
Formula:H₂O
MW:18.02 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
MDL Number:MFCD00011332
CAS Number:7732-18-5
Merck Index:12,10175
:Water
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Filtered through a 0.2 µm filter.
:Boiling Pt:
:Boiling Pt: 100 °C
:Melting Pt:
:Melting Pt: 0 °C
:Flash Pt:
:Flash Pt:
:M.W. 18.02 g/mol
EINECS:231-791-2
:100 °C
Storage Temperature:Ambient
:Filter 0,2 um
Specification Test Results
- Water for biotechnology nuclease-free, 0.2 µm filtered:
Specification Test Results
Customers Who Bought This Also Bought