Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3), Ultra Pure Grade
Catalog #: 97061-078
Supplier: VWR
CAS Number: 19817-92-6
Specifications
- Size:250 mg
- Cat. no.:97061-078
- Supplier No.:0145-250MG
Specifications
Specification Test Results
- Moisture (KF):= 13 %
- Purity (Dried):= 98 %
- Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3), Ultra Pure Grade:
Specification Test Results
About this item
Synonyms: Uridine-5'-triphosphoric acid trisodium salt
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 550.09 g/mol
Formula:C₉H₁₂N₂Na₃O₁₅P₃
MW:550.09 g/mol
MDL Number:MFCD00044310
CAS Number:19817-92-6
Customers Who Bought This Also Bought