Trypan blue 0.4% in aqueous solution ready-to-use, sterile
Catalog #: 97063-702
Supplier: VWR
Product available on GSA Advantage®
CAS Number: 72-57-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:100 mL
- Cat. no.:97063-702
- Supplier No.:K940-100ML
Specifications
Specification Test Results
- Abs.@ Lambda Max (1:100, Water):1 - 1.3
- Lambda Max (1:100, Water):585 nm - 595 nm
- Trypan blue 0.4% in aqueous solution ready-to-use, sterile:
Specification Test Results
About this item
Synonyms: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt,CI 23850,Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulfonic acid tetrasodium salt,3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt,Benzamine blue,Trypan blue sodium salt,Direct Blue 14,Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulfonate),AzidinBlue,3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulfonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt,Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 960.82 g/mol
:GHS08
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MW:960.82 g/mol
MDL Number:MFCD00003969
CAS Number:72-57-1
Customers Who Bought This Also Bought