L(+)-Aspartic acid ≥98%, powder, high purity
Supplier: VWR
Certificates
About this item
Synonyms: (S)-(+)-Aspartic acid,L(+)-2-Amino-1,4-butanedioic acid,(S)-(+)-2-Amino-1,4-butanedioic acid,H-Asp-OH,L-α-Aminosuccinic acid,L-αlpha-Aminosuccinic acid,(S)-2-Aminosuccinic acid,Asp
Formula:C₄H₇NO₄
MW:133.1 g/mol
Boiling Pt:311 °C (1013 hPa)
Melting Pt:251 °C
Density:1.6613 g/cm³ (13 °C)
MDL Number:MFCD00002616
CAS Number:56-84-8
Merck Index:13,00845
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Boiling Pt:
:Boiling Pt: 311 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 251 °C
:Flash Pt:
:Flash Pt:
:M.W. 133.1 g/mol
:L(+)-Aspartic acid
Storage Temperature:Ambient
EINECS:200-291-6
Specification Test Results
- Loss on Drying:= 0.2 %
- Purity:= 98 %
- L(+)-Aspartic acid ≥98%, powder, high purity:
Specification Test Results
Related Information
Customers Who Bought This Also Bought