L-Alanine, powder, high purity
Catalog #: 97061-024
Supplier: VWR
CAS Number: 56-41-7
Specifications
- Size:100 g
- Supplier No.:0106-100G
Specifications
Specification Test Results
- Arsenic:= 0.00015 %
- Ash:= 0.15 %
- Chloride:= 0.05 %
- Heavy Metals:= 0.001 %
- Sulfate:0.03 %
- Iron:0.003 %
- Loss on Drying:0.2 %
- pH (5%, Water) @25°C:5.5 - 7
- Purity:99 %
- Specific Rotation:13.7 -15.1 °
- L-Alanine, powder, high purity:
Specification Test Results
About this item
Synonyms: (S)-(+)-Alanine,H-Ala-OH,L(+)-Alanine,Alanine,L-α-Alanine,A,H-L-Ala-OH
Boiling Pt:189 °C (1013 hPa)
Melting Pt:314 °C…316 °C
Density:1.4 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 189 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 314 °C…316 °C
:Flash Pt:
:Flash Pt:
:M.W. 89.09 g/mol
Merck Index:13,00203
:L-Alanine
Formula:C₃H₇NO₂
MW:89.09 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00064410
CAS Number:56-41-7
EINECS:200-273-8
Customers Who Bought This Also Bought