Specifications
- Description:TRIS-GLYCINE BUFFER, 10X READY-PACK™
- Size:40 L
- Cat. no.:97063-782
- Application:Enough powder to make 40 L of 1X buffer. Each pack makes 1L of 10X concentrate. Add methanol to make standard transfer buffer.
- Supplier No.:M115-40L
Specifications
About this item
Following electrophoresis, proteins are frequently transferred from the polyacrylamide gel onto PVDF or nitrocellulose membranes for use in immunoblotting or sequencing
- Convenience of liquid concentrates or powder blend
- Consistent performance and quality
Although there are different methods by which to achieve transfer, the one most routinely used is electroblotting. Electroblotting utilizes conductive buffers that do not interfere with proteins binding to the membrane and can be performed in a vertical buffer tank or in a semi-dry blotting apparatus.