About this item
Blue BANDit™ is a safer and more environmentally friendly alternative to traditional Coomassie® Blue staining procedures.
- Ultra pure grade
- Sensitivity below 20 ng of protein per band
- No methanol or acetic acid—uses water for destaining
- Ready-to-use stain
This ready-to-use stain utilizes deionized water for destaining, thereby reducing the handling of hazardous materials and solvent waste in the lab. The easy protocol requires a pre-wash in water for 15 minutes, 1 hour of staining in Blue BANDit™, and a final water wash for 30 minutes. As the bands appear on the gel, the background remains clear, providing excellent visibility of proteins.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot NumberAvantor Top Rated Guide VWR™ Safe Planet Friendly Research Products
Product Literature
DFU-K217
Datasheet
VWR GHS Compliance Letter (US)