2-Propanol ≥70% in aqueous solution, AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: IPA,Isopropyl Rubbing Alcohol,Isopropyl alcohol,Dimethyl carbinol,2-Hydroxy propane,Isopropanol
Formula:(CH₃)₂CHOH
MW:60.1 g/mol
Density:0.8…0.92 g/cm³ (20 °C)
MDL Number:MFCD00011674
CAS Number:67-63-0
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:70
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: -89 °C
:Flash Pt:
:Flash Pt:
:M.W. 60.1 g/mol
:2-Propanol
EINECS:200-661-7
:-89
:82
UN:1219
:3
:II
:12
:GHS02
:GHS07
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Specific Gravity at 20°/20°C:0.872 - 0.883
- Acidity (mL):≤1.0
- Nonvolatile Residue:≤0.01 %
- Assay (by GC):68.0 - 72.0 %
- Appearance (clear, colorless liquid):
- in aqueous solution:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
sds_MACRV623
SDS
V623 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought