Methanol ≥99.5% (by GC), GenAR® NF, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Hydroxy methane,Carbinol,Wood alcohol
Formula:CH₃OH
MW:32.04 g/mol
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:12 °C
MDL Number:MFCD00004595
CAS Number:67-56-1
ADR:3,II
Merck Index:13,05984
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 64.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -98 °C|Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 12 °C
:M.W. 32.04 g/mol
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
REACH:01-2119433307-44
:3
:II
:12 °C
:GHS02
:GHS06
:GHS08
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets E.P. Chemical Specifications:
- Meets N.F. Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay (CH₃0H) (by GC):≥99.5 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Acidity:Passes Test
- NF - Alkalinity (as NH₃):≤3 ppm
- NF - Nonvolatile Residue:≤0.001 %
- NF - Readily Carbonizable Substances:Passes Test
- NF - Readily Oxidizable Substances:Passes Test
- NF - Acetone, Aldehydes (as (CH₃)₂CO):≤0.003 %
- NF - Water (H₂O):≤0.1 %
- EP - Identification A:Passes Test
- EP - Identification B:Passes Test
- EP/BP - Appearance:Passes Test
- EP - Acidity or Alkalinity:Passes Test
- EP - Relative Density:0.791 - 0.793
- EP - Absorbance at 230 nm (au):≤0.15
- EP - Absorbance at 250 nm (au):≤0.050
- EP - Absorbance at 270 nm (au):≤0.020
- EP - Absorbance at 290 nm (au):≤0.010
- EP - Scan from 230 nm to 290 nm:Passes Test
- EP/BP - Impurity A (Benzene):≤2 ppm
- EP - Related Substances - Single Impurity Peak:≤0.1 %
- EP - Related Substances - Total of All Impurity Peaks:≤0.3 %
- EP/BP - Reducing Substances:Passes Test
- EP/BP - Residue on Evaporation:≤10 ppm
- EP - Water:≤0.10 %
- Appearance (clear, colorless liquid):
- Only Class 3 Solvents are likely to be present. Residual concentration:
- of all Class 3 Solvents is <0.5%.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- Methanol ≥99.5% (by GC), GenAR® NF, Macron Fine Chemicals™:
Specification Test Results
Related Information
Certificates
Search by Lot Number8818 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
sds_MACR8818
SDS
spec_JT8818-14
Product specification
Customers Who Bought This Also Bought