EDTA dipotassium salt dihydrate, Ultrapure
Catalog #: 97062-274
Supplier: VWR
CAS Number: 25102-12-9
Specifications
- Packaging:HDPE bottle
- Size:500 g
- Cat. no.:97062-274
- Supplier No.:0720-500G
Specifications
Specification Test Results
- Chloride:= 0.005 %
- Heavy Metals:= 0.001 %
- Iron:= 0.001 %
- pH (5%, Water) @25°C:4.0 - 5.0
- Purity:= 99.0 %
- Appearance:White granular powder
- EDTA dipotassium salt dihydrate, Ultrapure:
Specification Test Results
About this item
Synonyms: Ethylenediaminetetra-acetic acid dipotassium salt dihydrate,Edetic acid dipotassium salt dihydrate,Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate,Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate,Potassium 2
Melting Pt:272 °C
Density:1 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 272 °C
:Flash Pt:
:Flash Pt:
:M.W. 404.46 g/mol
Merck Index:13,03546
Formula:C₁₀H₁₄K₂N₂O₈·2H₂O
MW:404.46 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00150036
CAS Number:25102-12-9
EINECS:217-895-0
Customers Who Bought This Also Bought