D-(-)-Ribose ≥98.0%, high purity
Catalog #: 97062-162
Supplier: VWR
CAS Number: 50-69-1
Specifications
- Packaging:HDPE bottle
- Size:100 g
- Cat. no.:97062-162
- Supplier No.:0671-100G
Specifications
Specification Test Results
- Arsenic:0.0002 %
- Ash:= 0.2 %
- Heavy Metals (as Pb):0.001 %
- Loss on Drying:= 3.0 %
- Purity:98.0 %
- Specific Rotation (4%, Water):(-21.5°) to (-19.7°)
- D-(-)-Ribose ≥98.0%, high purity:
Specification Test Results
About this item
Synonyms: (2R,3R,4R)-2,3,4
Boiling Pt:331 °C (1013 hPa)
Melting Pt:90…95 °C
Density:0.8 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0
:Boiling Pt:
:Boiling Pt: 331 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 90…95 °C
:Flash Pt:
:Flash Pt:
:M.W. 150.13 g/mol
Merck Index:12,08371
Formula:C₅H₁₀O₅
MW:150.13 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00135453
CAS Number:50-69-1
EINECS:200-059-4
Customers Who Bought This Also Bought