DL-Malic acid 99-100.5%, high purity
Catalog #: 97062-140
Supplier: VWR
CAS Number: 617-48-1
Specifications
- Packaging:HDPE bottle
- Size:2.5 kg
- Cat. no.:97062-140
- Supplier No.:0664-2.5KG
Specifications
Specification Test Results
- Fumaric Acid:1%
- Heavy Metals (as Pb):= 0.001%
- Insolubles:= 0.1%
- Maleic Acid:= 0.05%
- Purity:99% - 100.5%
- Residue on Ignition:= 0.1%
- DL-Malic acid 99-100.5%, high purity:
Specification Test Results
About this item
Synonyms: DL-Hydroxysuccinic acid,(±)-Malic acid,(±)-Hydroxy-1,4-butanedioic acid,(±)-Hydroxysuccinic acid,Hydroxybutanedioic acid
:DL-Malic acid
Boiling Pt:306 °C (1013 hPa)
Melting Pt:131…132 °C
Density:1.601 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99-100,5
:Boiling Pt:
:Boiling Pt: 306 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 131…132 °C
:Flash Pt:
:Flash Pt:
:M.W. 134.09 g/mol
Merck Index:13,05730
MW:134.09 g/mol
MDL Number:MFCD00064212
EINECS:210-514-9
:GHS05
:GHS07
Formula:HOOCCH(OH)CH₂COOH
Storage Temperature:Ambient
CAS Number:617-48-1
Customers Who Bought This Also Bought