D-(+)-Glucose monohydrate
Supplier: VWR
Certificates
About this item
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate
CAS Number:14431-43-7
:5996-10-1
Formula:C₆H₁₂O₆·H₂O
MW:198.17 g/mol
MDL Number:MFCD00149450
:D-(+)-Glucose monohydrate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 83 °C
:Flash Pt:
:Flash Pt:
:M.W. 198.17 g/mol
:83
Specification Test Results
- DNase:none detected
- Insolubles:= 0.005 %
- Loss on Drying:8 % - 9 %
- Purity (Anhydrous):= 99.5 %
- RNase:none detected
- Specific Rotation:52.6 °- 53.2°
- D-(+)-Glucose monohydrate:
Specification Test Results
Customers Who Bought This Also Bought