Specifications
- Description:Hematology analyzer slide
- Supplier No.:EMS100TPB
Specifications
About this item
Azer Scientific's Hematology Analyzer Slides are designed for use in automated hematology slide makers.
- Pure white glass
- Two white, printed bars to prevent jamming
- Beckman Coulter® systems compatible
- 75 x 25 x 1 mm
- Clipped corners
Manufactured from 100% pure white glass for the highest quality in hematology applications, these slides also feature two printed bars to help prevent jamming in equipment.
They are compatible with Beckman Coulter® systems.